Learn More
Cayman Chemical Psoala coylfola Bavacn 10mg
SDP
Supplier:  Cayman Chemical 1168510 MG
Encompass_Preferred
A phytoestrogen that activates ERα and ERβ (EC50s = 320 and 680 nM, respectively); stimulates osteoblast proliferation and differentiation and prevents bone loss following ovariectomy in rats
Catalog No. 50-295-2195