Learn More
Cayman Chemical Psorala corylfola nglcn 10mg
SDP
Supplier:  Cayman Chemical 1168310 MG
Encompass_Preferred
A furanocoumarin with antibacterial activities; prevents tacrine-induced cytotoxicity in HepG2 cells (EC50 = 47 μg/ml) and vascular relaxation in phenylephrine-precontracted rat aorta; weakly inhibits topoisomerase II (IC50 = 404 μM)
Catalog No. 50-295-1806