Learn More
Julabo USA Inc Each, Pt100 sensor 6x300mm, 1.5m cable, SS
SDP
Supplier:  Julabo USA Inc 8981010
Encompass_Preferred
Bath Accessories, Julabo, External Pt100 sensor, 300 x 6 mm, diameter, stainless steel, 1.5 m cable with 4-pin Lemosa plug. For use with JULABO systems having external Pt100 capability.
Catalog No. 50-753-4121