Learn More
Selleck Chemical LLC PT-179-E1937-1G
SDP
Supplier:  Selleck Chemical LLC E19371G
Encompass_Preferred
PT-179 an orthogonal Thalidomide derivative targets cereblon without causing off-target degradation effects PT-179 specifically binds CRBN forms a ternary complex with a target protein fused to a zinc finger (ZF) degron and mediates the degradation of the tagged protein
Catalog No. 50-313-7843