Learn More
Medchemexpress LLC Pt(II) protoporphyrin IX | 98303-94-7 | 80.0% | 755.73 g/mol | C34H32N4O4Pt | 5 MG

Supplier: Medchemexpress LLC HY136476G5MG
Pt(II) protoporphyrin IX is a platinum(II)-metalated derivative of protoporphyrin IX used as a research reagent and chemical intermediate. It is a tetrapyrrolic heterocycle related to heme biosynthesis, with molecular formula C34H32N4O4 and molecular weight 755.73 g/mol. Typical purity is about 80.0% and the material is stored as a powder at low temperatures for long-term stability.
- Platinum(II)-metalated protoporphyrin core for mechanistic and spectroscopic studies.
- Suitable as a chemical intermediate and research reagent.
- Molecular formula C34H32N4O4 and molecular weight 755.73 g/mol for accurate dosing.
- Typical purity approximately 80.0% for experimental planning.
- Powder form with recommended storage at -20°C (powder) and colder for solutions to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.