Learn More
Glass Expansion Platinum Skimmer Cone for Thermo Scientific Q/RQ/TQ Series ICP-MS (Insert-free cone replaces combination of 1324540 & 1318480 (3.5mm High Matrix Insert); 1324540 & 1311880 (2.8mm High Sensitivity Insert)
SDP
Supplier:  Glass Expansion TG1048PT
Encompass_Preferred
Platinum Skimmer Cone, Cold and Hot Plasma (1341430) ¿ Suitable for Thermo Q/RQ/TQ ICP-MS. specifically designed to operate ONLY with the Cold Plasma Lens Kit (1341380). This skimmer cone can be used with the Cold Plasma Lens Kit to operate in both Cold (RF Power 550 W) and Hot Plasma conditions (RF Power 1550 W). Although the highest cost option compared to Ni, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1028) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For high matrix tolerance and high sensitivity, we recommend TG1048-Pt.For Robust plasma conditions, we recommend TG1045-Ni.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2931