Learn More
XRF Scientific Americas Inc. PLATINUM DISH SEMI-ROUND SHAPE 250ML FROM XRF SCIENTIFIC AMERICAS
Supplier:  XRF Scientific Americas Inc. GD250
Encompass
94x42mm with a 250 ml capacity. Lids sold separately. These 99.95% pure platinum dishes are available in semi-round shape with flat base, & used for Fusions, Digestion, Ash testing, Loss on Ignition (LOI). Due to their high temperature strenght, ductility, corrosion and oxidation, excellent resistance to chemical attacks by acids and fusion mixtures resistance; these platinum crucibles are critical when high temperatures & resistance to chemical attacks are required.
Catalog No. NC1231710
Only null left
Add to Cart
Request a Quote