Learn More
Sigma Aldrich Fine Chemicals Biosciences PTAA a polytriaryl amine s1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 702471
Encompass
PTAA poly(triaryl amine) semiconductor is an organic p-type semiconductor with hole mobilities of 10A3nbspup to 10A2nbspcm2nbspVA1nbspsA1 which results in a high carrier mobility. It is a stable glassy polymer and has good ionization potential for thick film diodes.
Catalog No. 50-290-6854