Learn More
Apexbio Technology LLC PTC-209, 5mg. Cas: 315704-66-6 MFCD: MFCD02110649. Bmi-1 inhibitor,cell-permeable and small-molecule
SDP
Supplier:  Apexbio Technology LLC B31795
Encompass_Preferred
PTC-209 is a small-molecule inhibitor of BMI-1 with IC50 value of 0.5 M [1].PTC-209 is screened out by using gene expression modulation by small molecules (GEMS) technology. It can lower BMI-1 transcript levels. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5343