Learn More
Selleck Chemical LLC PTC-209 HBr
SDP
Supplier:  Selleck Chemical LLC S753925MG
Encompass_Preferred
PTC-209 HBr is the hydrobromide salt of PTC-209 which is a potent and selective BMI-1 inhibitor with IC50 of 0 5 M and results in irreversible reduction of cancer-initiating cells (CICs)
Catalog No. 50-000-04569