Learn More
Selleck Chemical LLC PTC-209 HBr 5mg 1217022-63-3
SDP
Supplier:  Selleck Chemical LLC S75395MG
Encompass_Preferred
PTC-209 HBr is the hydrobromide salt of PTC-209, which is a potent and selective BMI-1 inhibitor with IC50 of 0.5 ?M, and results in irreversible reduction of cancer-initiating cells (CICs). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4918