Learn More
Medchemexpress LLC Ptda-peg4-azide | 2639395-40-5 | 539.54 g/mol | C22H33N7O9 | 50 MG

Supplier: Medchemexpress LLC HY13094050MG
PTDA-PEG4-azide is a PEG4-based azide linker used in click chemistry and antibody-drug conjugate (ADC) synthesis. It provides an azide functional group compatible with copper-catalyzed and strain-promoted cycloaddition reactions.
- Contains azide functional group.
- Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
- Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN reagents.
- Peg4 spacer offers flexibility and improved solubility in conjugates.
- Molecular weight approximately 539.54 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.