Learn More
Cayman Chemical PtdIns 3 4 5P31 2dIpalm 100UG
SDP
Supplier:  Cayman Chemical 64920100 UG
Encompass_Preferred
A synthetic analog of natural PtdIns featuring C16:0 fatty acids at the sn-1 and sn-2 positions; contains the same inositol and DAG stereochemistry as the natural compound
Catalog No. 50-289-6957