Learn More
Cayman Chemical PtdIns 3 4 5P31 2dIpalm 100UG
SDP
Supplier:  Cayman Chemical 9000414100 UG
Encompass_Preferred
An internal standard for the quantification of PtdIns-(3,4,5)-P3 (1,2-dipalmitoyl) by GC- or LC-MS
Catalog No. 50-289-6222