Learn More
Cayman Chemical PtdIns 3 4 5P3biotin sodium
SDP
Supplier:  Cayman Chemical 1000953125 UG
Encompass_Preferred
PtdIns-(3,4,5)-P3-biotin is an affinity probe which allows the PIP3 to be detected through an interaction with the biotin ligand. This design allows PtdIns-(3,4,5)-P3-biotin to serve as a general probe for any protein with a high affinity binding interaction with inositol-(3,4,5)-triphosphate phospholipids, such as phosphatidylinositol 3-kinase, PTEN, or PH-domain-containing proteins.
Catalog No. 50-288-9431