Learn More
Cayman Chemical PtdIns 3P1 1 2dipalmitoyld62
SDP
Supplier:  Cayman Chemical 10005616100 UG
Encompass_Preferred
An internal standard for the quantification of PtdIns-(3)-P1 (1,2-dipalmitoyl) (ammonium salt) by GC- or LC-MS.
Catalog No. 50-288-6487