Learn More
Cayman Chemical PtdIns 4 5P2 1 2dipalmitoyld
SDP
Supplier:  Cayman Chemical 10005615100 UG
Encompass_Preferred
An internal standard for the quantification of PtdIns-(4,5)-P2 (1,2-dipalmitoyl) (sodium salt) by GC- or LC-MS.
Catalog No. 50-288-6926