Learn More
Medchemexpress LLC Pterin | 2236-60-4 | MFCD01456481 | 99.8% | 163.14 g·mol⁻¹ | C6H5N5O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW037825
Encompass_Preferred
Pterin is a pteridine heterocycle provided as a high-purity research reagent for biochemical and medicinal chemistry applications.
  • High purity (99.75%) for analytical and synthetic work.
  • Molecular formula C6H5N5O and molecular weight 163.14 g·mol⁻¹.
  • Available in small to mid scale pack sizes (25 mg-5 g) for flexible usage.
  • Characterized by supporting data including COA, HNMR, LC-MS, and HPLC.
  • Suitable as a reference standard or building block in research workflows.
Catalog No. 50-003-03849