Learn More
Cytiva (Formerly Pall Lab) wwPTFE Membrane Disc Filter - 0.2 m, 25 mm (50/pkg)
Supplier:  Cytiva 60538
Encompass
Membrane with Chemical Compatibility for Aqueous Solutions and Aggressive Solvents-Maximum versatility. Filters aqueous solutions or aggressive chemicals. -Low protein binding membrane gives high recovery of critical proteinaceous samples.-HPLC certified. Provides assurance that the filter will not add artifacts to your analysis.-Low API adsorption. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3252