Learn More
Chemglass Life Sciences PTFE 1 in Male NPT Plug for 1 in NPT Ports
SDP
Supplier:  Chemglass Life Sciences CG1945T89
Encompass_Preferred
This PTFE 1 in male NPT plug provides a chemically inert, corrosion-resistant closure for 1 in NPT ports and threaded fittings. It is suitable for laboratory and general industrial use where a non-reactive, leak-resistant seal is required. Sold individually.
  • Made from PTFE for broad chemical compatibility and low permeability.
  • 1 in male NPT thread for sealing standard 1 in NPT ports.
  • Sold individually (each) for single-part replacement or spares.
  • Provides a reliable, leak-resistant closure for temporary or permanent sealing.
  • Suitable for laboratory and general industrial applications.
Catalog No. NC3850712