Learn More
Ace Glass, Inc. Adapter, filter, PTFE, #25 Ace-Thred to 1/4in-28UNF, 100 micron polyethylene filter, FETFE O-ring
SDP
Supplier:  Ace Glass, Inc. 583851
Encompass_Preferred
Adapter, filter, PTFE, #25 Ace-Thred to 1/4in-28UNF, 100 micron polyethylene filter, FETFE O-ring
Catalog No. NC1823837