Learn More
Chemglass Life Sciences Chemglass PTFE compression fitting, 1 in (for heavy-wall PTFE tubing)
SDP
Supplier:  Chemglass Life Sciences CG1975100
Encompass_Preferred
Chemglass CG-1975-100 is a 1 in PTFE compression fitting designed to provide a chemically inert connection for heavy-wall PTFE tubing used in process reactors and laboratory chemistry assemblies. The fitting and ferrules are machined from virgin PTFE for chemical resistance, while grippers and nuts are made from PVDF to ensure secure, corrosion-resistant connections.
  • Provides chemically inert PTFE construction for broad chemical resistance.
  • Compatible with heavy-wall PTFE tubing for secure fluid transfer.
  • PVDF grippers and molded nuts for durable, corrosion-resistant holding.
  • Designed for use in process reactors and laboratory chemistry equipment.
  • Low leach and contamination risk suitable for sensitive chemistries.
  • Easy to install and assemble for routine maintenance.
Catalog No. NC2607638