Learn More
Perkin Elmer US LLC PTFE Flame Sampling Probe Assembly
Supplier:  Perkin Elmer US LLC N2024131
Encompass_Preferred
PTFE probes provide laminar flow and are the most resistant to chemical degradation.
Catalog No. 50-211-1644