Learn More
Ace Glass, Inc. PTFE SEAL TAPE 1/2IN PTFE SEAL TAPE 1/2IN
SDP
Supplier:  Ace Glass, Inc. 1412018
Encompass
99% pure PTFE tape used to create a leak-tight seal when connecting items such as a pressure gauge to an adapter. Wrap tape around male thread before threading into female. Thickness is .0035".
Catalog No. NC9060869