Learn More
Sigma Aldrich Fine Chemicals Biosciences PTfE sealing washers CGA I
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1469941PAK
Encompass
Use with Aldrich lecture-bottle regulators and control valves.
Catalog No. 50-289-9999