Learn More
Cole-Parmer PTFE Tissue Grinder, Plain Plunger, 50 mL, 270 mm L
Supplier:  Cole-Parmer 4446806
Encompass_Preferred
Plain tip plungers effectively homogenize a variety of tissue samples. Capable of reducing cellualar tissue to primary cells. Plunger head is pure PTFE with 6.5 mm stainless steel shaft.
Catalog No. NC1973392