Learn More
Ace Glass, Inc. PTFE tubing, 2.0mm id, 0.3mm wall, 3 meters
SDP
Supplier:  Ace Glass, Inc. 1268427
Encompass
PTFE tubing,2.0mm id,0.3mm wall,3 metersClear, small diameter, PTFE tubing for many Chromatography needs. Use our 5855 and 5838 series connectors. 3 meter lengths.
Catalog No. NC3256650