Learn More
Medchemexpress LLC HY-122716 5mg Medchemexpress, PTGR2-IN-1 CAS:349093-44-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1227165MG
Encompass_Preferred
Medchemexpress, HY-122716 5mg PTGR2-IN-1 CAS:349093-44-3 PTGR2-IN-1 is a potent PTGR2 inhibitor with an IC50 of ~0.7 μM. PTGR2-IN-1 increases 15-keto-PGE2-dependent PPARγ transcriptional activity in PTGR2-transfected HEK293T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0992