Learn More
Medchemexpress LLC pTH (1-37) (human) | 136799-54-7 | 99.9% | 4401.08 | 1 MG
Supplier: Medchemexpress LLC HYP48231MG
pTH (1-37) (human) is a fragment of parathyroid hormone (PTH) that induces cAMP formation and increases alkaline phosphatase activity. It has been shown to increase growth, bone calcium content, and bone mineral density (BMD) in uremic animals.
- Induces cAMP formation and increases alkaline phosphatase activity.
- Increases growth, bone calcium content, and bone mineral density (BMD) in uremic animals.
- Has potential for the research of osteoporosis.