Learn More
Medchemexpress LLC Pth-related protein (1-34) (human,mouse,rat) | 112540-82-6 | 98.7% | 4018.00 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP30751MG
Encompass_Preferred
Pth-related protein (1-34) (human, mouse, rat) is a fragment of parathyroid hormone-related protein. It acts as an anabolic agent and is utilized in studies related to postmenopausal osteoporosis. This product is intended for research purposes only and is not for direct patient use.
  • Fragment of parathyroid hormone-related protein
  • Anabolic agent
  • Used for postmenopausal osteoporosis study
  • For research use only
Catalog No. 50-003-96655