Learn More
Medchemexpress LLC PTH-tyrosine | 4332-95-0 | MFCD00027415 | ≥98.0% | 298.36 g/mol | C16H14N2O2S | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW282259250MG
Encompass_Preferred
PTH-tyrosine is a phenylthiohydantoin-derivatized tyrosine supplied for biochemical and analytical research. As a small-molecule reagent (C16H14N2O2S, MW 298.36), it is used in peptide sequencing workflows and other assays that require a derivatized amino-acid intermediate. The product is offered in multiple laboratory-scale package sizes and is accompanied by lot-specific analytical documentation where available.
  • Used in biochemical assays and peptide analysis.
  • Phenylthiohydantoin derivative with defined molecular formula and weight.
  • Suitable for analytical workflows requiring derivatized amino acids.
  • Available in multiple laboratory-scale package sizes for flexibility.
  • Certificate of analysis available for lot-specific analytical data.
Catalog No. 50-002-81316