Learn More
Medchemexpress LLC PTH-tyrosine | 4332-95-0 | MFCD00027415 | >98.0% | 298.36 g/mol | C16H14N2O2S | 5 G
SDP
Supplier:  Medchemexpress LLC HYW2822595G
Encompass_Preferred
PTH-tyrosine is the phenylthiohydantoin derivative of tyrosine used as a biochemical reagent and reference standard in peptide sequencing and analytical assay workflows. Supplied as a characterized solid, it is suitable for laboratory analytical applications and is provided with documentation (datasheet/COA) detailing storage and handling requirements.
  • Phenylthiohydantoin derivative of tyrosine, useful as a sequencing reagent.
  • Characterized by CAS 4332-95-0 and formula C16H14N2O2S.
  • Molecular weight approximately 298.36 g/mol.
  • High purity commonly ≥98% by HPLC, suitable for analytical use.
  • Available in multiple pack sizes to support laboratory workflows.
  • Store under conditions specified in the certificate of analysis.
Catalog No. 50-002-82317