Learn More
Eurofins Discoverx ChemiScreen PTH2 Parathyroid Hormone Receptor Stable Cell Line, 2 vials. (HTS173C)
Supplier:  Eurofins Discoverx HTS173C
Encompass
The ChemiScreen PTH2 Parathyroid Hormone Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. parathyroid hormone) based activation of PTH2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTH2R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of PTH2R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-6003
Only null left
Add to Cart
Request a Quote