Learn More
Medchemexpress LLC PTP inhibitor 1 | 2632-13-5 | 99.55% | 229.07 | 50 G

Supplier: Medchemexpress LLC HYW01347850G
PTP inhibitor 1 is a protein tyrosine phosphatase (PTP) inhibitor that has an anti-angiogenic effect. It is for research use only and not sold to patients.
- Is a protein tyrosine phosphatase (PTP) inhibitor.
- Exhibits anti-angiogenic activity with an IC50 of 3.7 μM for HUVECs.
- Available in solid form, off-white to light yellow.
- Soluble in DMSO (≥ 100 mg/mL).
- Storage: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 2 years, -20°C for 1 year.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.