Learn More
Medchemexpress LLC PTP inhibitor 1 | 2632-13-5 | 99.6% | 229.07 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01347810MM/1ML
Encompass_Preferred
PTP inhibitor 1 is a protein tyrosine phosphatase (PTP) inhibitor, with anti-angiogenic effect. It shows anti-angiogenic activity with an IC50 of 3.7 μM for HUVECs.
  • Protein tyrosine phosphatase (PTP) inhibitor
  • Anti-angiogenic effect
Catalog No. 50-004-54683