Learn More
TARGETMOL CHEMICALS INC PTP INHIBITOR IV 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T221375MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PTP Inhibitor IV is an effective protein tyrosine phosphatase (PTP) inhibitor which can selectively inhibit DUSP14 phosphatase activity (IC50.5.21uM). PTP Inhibitor IV inhibited SHP-2 PTP1B PTp-(epsilon) PTP Meg-2 PTP-(sigma) PTP-(beta) and PTP-u IC50 were 1.8 uM 2.5 uM 8.4 uM 13 uM 20 uM 6.4 uM and 6.7 uM respectively. PTP Inhibitor IV affects cellular signaling by inhibiting PTP activity and may regulate specific pathways associated with PTP-mediated signaling events. purity: 99%
Catalog No. 50-258-0976