Learn More
Cayman Chemical PTP InhIbItr I 100mg
SDP
Supplier:  Cayman Chemical 19766100 MG
Encompass_Preferred
A SHP-1 and PTP1B inhibitor (Kis = 43 and 42 µM for the SHP-1 catalytic domain and full-length PTP1B, respectively); inhibits endothelial cell network formation of HUVECs cultured with normal human dermal fibroblasts (IC50 = 3.7 µM); has been used in the synthesis of compounds with antibacterial, antifungal, fungicidal, and anticancer activities
Catalog No. 50-298-2052