Learn More
Cayman Chemical PTP InhIbItr III 1mg
SDP
Supplier:  Cayman Chemical 190591 MG
Encompass_Preferred
A photoreversible covalent inhibitor of PTPs; binds the catalytic domain of SHP-1 (Ki = 184 µM) and covalently reacts with free thiols, a reaction that is reversible with irradiation (350 nm)
Catalog No. 50-298-1940