Learn More
Medchemexpress LLC HY-W054146 50mg Medchemexpress, PTP1B-IN-9 CAS:145888-79-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW05414650MG
Encompass_Preferred
Medchemexpress, HY-W054146 50mg PTP1B-IN-9 CAS:145888-79-5 PTP1B-IN-9 is a ubiquitin-proteasome system (UPS)-stressor. PTP1B-IN-9 inhibits ubiquitin-mediated protein degradation upstream of the 20S proteasomal catalytic activites. PTP1B-IN-9 triggers a ubiquitin-proteasome-system (UPS)-stress response without affecting 20S proteasome catalytic activities. Anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3492