Learn More
Aviva Systems Biology Corp HACD4 Antibody - middle region (ARP44598_P050) | 100uL
Supplier: Aviva Systems Biology Corp ARP44598_P050
This is a rabbit polyclonal antibody against PTPLAD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTPLAD2. Alternate names: DKFZp686F01145, DKFZp686G24132, HACD4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat