Learn More
TARGETMOL CHEMICALS INC PU.1-IN-1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7358510MG
Encompass_Preferred
Also available in 25 mg 50 mg and bulk. Please contact Fisher for quotes. PU.1-IN-1 is a potent PU.1 inhibitor (IC50 2 nM) with anti-inflammatory activity. purity: 98%
Catalog No. 50-257-9205