Learn More
Medchemexpress LLC PU-H54 | 1454619-13-6 | 99.0% | 337.44 g/mol | C18H19N5S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11739550MG
Encompass_Preferred
PU-H54 is a Grp94-selective Hsp90 inhibitor supplied as a solid (white to off-white) for research use in biochemical and cellular studies. It is provided with analytical purity and recommended storage conditions for both powder and solution forms.
  • Grp94-selective Hsp90 inhibitor for biochemical and cellular studies.
  • High purity, 99.0%, suitable for research applications.
  • Solid, white to off-white appearance for straightforward handling.
  • Available in multiple pack sizes, including a 50 mg quantity.
  • Stability data provided for powder and in-solution storage conditions.
Catalog No. 50-002-28642