Learn More
Medchemexpress LLC HY-124696 5mg , PU139 CAS:158093-65-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246965MG
Encompass_Preferred
HY-124696 5mg PU139 CAS: 158093-65-3 PU139 is a potent pan-histone acetyltransferase (HAT) inhibitor. PU139 blocks the HATs Gcn5, p300/CBP-associated factor (PCAF), CREB (cAMP response element-binding) protein (CBP) and p300 with IC50s of 8.39, 9.74, 2.49 and 5.35 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9661