Learn More
ATCC Puccinia canaliculata (Schweinitz) Lagerheim, teleomorph - ATCC 40199 (strain Tifton)
Supplier: ATCC 40199
Puccinia canaliculata (Schweinitz) Lagerheim, teleomorph (strain Tifton) is a rust fungus supplied for agricultural and plant pathology research, including studies of biological control of yellow nutsedge (Cyperus esculentus). This material is provided for research use; viability testing and warranty apply when handled according to the provider's instructions.
- Used for agricultural and plant pathology research on yellow nutsedge (Cyperus esculentus).
- Isolated strain Tifton.
- Supplied as a fungal culture intended for laboratory research.
- Store in the vapor phase of liquid nitrogen for long-term preservation.
- Handled at biosafety level 1 (BSL 1).
- Viability is warranted for 30 days from shipment when handled according to the provider's instructions.