Learn More
Medchemexpress LLC HY-N0145 10mg Medchemexpress, Puerarin CAS:3681-99-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN014510MG
Encompass_Preferred
Medchemexpress, HY-N0145 10mg Puerarin CAS:3681-99-0 Puerarin, an isoflavone extracted from Radix puerariae, is a 5-HT2C receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3424