Learn More
Medchemexpress LLC Pulcherriminic acid | 957-86-8 | 256.30 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN104731MG
Encompass_Preferred
Pulcherriminic acid is a cyclic dipeptide antimicrobial agent with high affinity for Fe3+, found mainly in Bacillus and yeast. It chelates iron ions through a non-enzymatic reaction to form the extracellular red pigment pulcherrimin, which competes for iron nutrition and thus achieves an antibacterial effect. Pulcherriminic acid has applications in food, agriculture and medical industries.
  • Can inhibit the formation of Bacillus subtilis biofilm.
  • Can form insoluble red pigment pulcherrimin in the presence of Fe3+ to compete for iron nutrients, thereby inhibiting the growth of Metschnikowia yeasts.
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Cyclopeptides
  • Initial source: Microorganisms (Bacillus and yeast)
Catalog No. 50-004-16939