Learn More
Medchemexpress LLC Pulcherriminic acid | 957-86-8 | 256.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN104735MG
Encompass_Preferred
Pulcherriminic acid is a cyclic dipeptide antimicrobial agent with high affinity for Fe3+, found mainly in Bacillus and yeast. It chelates iron ions through a non-enzymatic reaction to form the extracellular red pigment pulcherrimin, which competes for iron nutrition and thus achieves an antibacterial effect. It has applications in food, agriculture and medical industries.
  • Cyclic dipeptide antimicrobial agent
  • Exhibits high affinity for Fe3+
  • Chelates iron ions to form extracellular red pigment pulcherrimin
  • Competes for iron nutrition, achieving an antibacterial effect
  • Applicable in food, agriculture, and medical industries
  • Inhibits Bacillus subtilis biofilm formation
  • Inhibits Metschnikowia yeasts growth by rapid iron removal
Catalog No. 50-004-15272