Learn More
Medchemexpress LLC Pulcherriminic acid | 957-86-8 | 98.0% | 256.30 g·mol⁻1 | C12H20N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN1047310MG
Encompass_Preferred
Pulcherriminic acid is a cyclic dipeptide iron chelator that binds Fe3+ to form the extracellular pigment pulcherrimin. It is used in biochemical and microbiology research to study iron metabolism, microbial competition, and antimicrobial activity.
  • High affinity for Fe3+ for iron chelation studies.
  • Cyclic dipeptide structure relevant to pigment formation research.
  • Purity 98.0% suitable for biochemical assays.
  • Light yellow solid for straightforward handling and storage.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-96076