Learn More
Medchemexpress LLC Pulegone | 89-82-7 | 152.24 | 1 G

Supplier: Medchemexpress LLC HYN15001G
Pulegone is a monoterpene ketone compound found extensively in the essential oils of various plants. It serves as a bird repellent and exhibits diverse activities including anti-inflammatory, antibacterial, antifungal, and anti-hyperalgesic effects. It is particularly effective against bacteria of the Salmonella species.
- Monoterpene ketone compound
- Widely present in essential oils of many plants
- Purity: 99.57%
- Appearance: Liquid (Density: 0.937 g/cm3)
- Color: Colorless to light yellow
- Bird repellent
- Anti-inflammatory agent
- Antibacterial agent, particularly against Salmonella species
- Antifungal agent
- Anti-hyperalgesic effects
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.