Learn More
Medchemexpress LLC Pulegone | 89-82-7 | 152.24 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN150010MM/1ML
Encompass_Preferred
Pulegone is a monoterpene ketone compound found in the essential oils of many plants. It is known for its anti-inflammatory, antibacterial, antifungal, and anti-hyperalgesic properties, and can also be used as a bird repellent. Pulegone is particularly effective against bacteria of the Salmonella species.
  • Inhibits inflammatory mediators by downregulating iNOS, COX-2, NF-κB, and MAPK signaling pathways.
  • Upregulates Nrf-2/HO-1.
  • Increases intracellular calcium concentration in chicken sensory neurons.
  • Activates TRPM8 and TRPA1 channels.
  • Shows beneficial anti-hyperalgesic effects in inflammatory pain models.
Catalog No. 50-004-16455